missing translation for 'onlineSavingsMsg'
Learn More

Niobium potassium isopropoxide, 2.5% w/v in isopropanol, 99% (metals basis), Thermo Scientific™

Product Code. 11363538
Change view
Click to view available options
Quantity:
50 mL
200 mL
Unit Size:
200mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11363538 200 mL 200mL
11353538 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11363538 Supplier Thermo Scientific Alfa Aesar Supplier No. 039165.AH
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21864-23-3
Molecular Formula C12H31KNbO4
Molecular Weight (g/mol) 371.381
MDL Number MFCD00798516
InChI Key WLSOPSYOUMIABE-UHFFFAOYSA-N
Synonym niobium potassium isopropoxide w/v in isopropanol
PubChem CID 121233692
IUPAC Name potassium;niobium;propan-2-ol;propan-2-olate
SMILES CC(C)O.CC(C)O.CC(C)O.CC(C)[O-].[K+].[Nb]

Specifications

Color Brown to Yellow
Flash Point 12°C (54°F)
Odor Alcohol-like
Linear Formula KNb[OCH(CH3)2]6
Quantity 200 mL
UN Number UN1219
Sensitivity Moisture sensitive
Formula Weight 486.53
Percent Purity 99%
Concentration or Composition (by Analyte or Components) 2.5% w/v in isopropanol
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Niobium potassium isopropoxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.