accessibility menu, dialog, popup

UserName

CH3CH3HOCH3CH3HOCH3CH3HOCH3CH3OKNb

CAS RN 21864-23-3

IUPAC Name: potassium tris(propan-2-ol) niobium propan-2-olate
Synonyms: potassium tris(isopropyl alcohol) niobium propan-2-olate
Molecular Weight (g/mol): 371.38
Molecular Formula: C12H31KNbO4
InChi Key: WLSOPSYOUMIABE-UHFFFAOYSA-N
SMILES: [K+].[Nb].CC(C)O.CC(C)O.CC(C)O.CC(C)[O-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results