missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicotinic Acetylcholine Receptor beta Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£157.00 - £373.00
Specifications
| Antigen | Nicotinic Acetylcholine Receptor beta |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Nicotinic Acetylcholine Receptor beta Polyclonal antibody specifically detects Nicotinic Acetylcholine Receptor beta in Human, Mouse samples. It is validated for Western BlotSpecifications
| Nicotinic Acetylcholine Receptor beta | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Alzheimers Research, Neurodegeneration, Neuroscience | |
| PBS (pH 7.3), 50% glycerol | |
| 1140 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| acetylcholine receptor subunit beta, ACHRBSCCMS, cholinergic receptor, nicotinic, beta 1 (muscle), cholinergic receptor, nicotinic, beta polypeptide 1 (muscle), CHRNB, CMS1D, CMS2A, FLJ57107 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 25-235 of human CHRNB1 (NP_000738.2). EAEGRLREKLFSGYDSSVRPAREVGDRVRVSVGLILAQLISLNEKDEEMSTKVYLDLEWTDYRLSWDPAEHDGIDSLRITAESVWLPDVVLLNNNDGNFDVALDISVVVSSDGSVRWQPPGIYRSSCSIQVTYFPFDWQNCTMVFSSYSYDSSEVSLQTGLGPDGQGHQEIHIHEGTFIENGQWEIIHKPSRLIQPPGDPRGGREGQRQEV | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title