missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicotinic Acetylcholine Receptor beta Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93445-0.1ml
This item is not returnable.
View return policy
Description
Nicotinic Acetylcholine Receptor beta Polyclonal antibody specifically detects Nicotinic Acetylcholine Receptor beta in Human, Mouse samples. It is validated for Western Blot
Specifications
| Nicotinic Acetylcholine Receptor beta | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| acetylcholine receptor subunit beta, ACHRBSCCMS, cholinergic receptor, nicotinic, beta 1 (muscle), cholinergic receptor, nicotinic, beta polypeptide 1 (muscle), CHRNB, CMS1D, CMS2A, FLJ57107 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 25-235 of human CHRNB1 (NP_000738.2). EAEGRLREKLFSGYDSSVRPAREVGDRVRVSVGLILAQLISLNEKDEEMSTKVYLDLEWTDYRLSWDPAEHDGIDSLRITAESVWLPDVVLLNNNDGNFDVALDISVVVSSDGSVRWQPPGIYRSSCSIQVTYFPFDWQNCTMVFSSYSYDSSEVSLQTGLGPDGQGHQEIHIHEGTFIENGQWEIIHKPSRLIQPPGDPRGGREGQRQEV | |
| 0.1 mL | |
| Alzheimers Research, Neurodegeneration, Neuroscience | |
| 1140 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction