missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicotinic Acetylcholine R alpha 2/CHRNA2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£149.00 - £366.00
Specifications
| Antigen | Nicotinic Acetylcholine R alpha 2/CHRNA2 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18615621
|
Novus Biologicals
NBP2-94552-0.02ml |
0.02 mL |
£149.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18642761
|
Novus Biologicals
NBP2-94552-0.1ml |
0.1 mL |
£366.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nicotinic Acetylcholine R alpha 2/CHRNA2 Polyclonal antibody specifically detects Nicotinic Acetylcholine R alpha 2/CHRNA2 in Rat samples. It is validated for Western BlotSpecifications
| Nicotinic Acetylcholine R alpha 2/CHRNA2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Alzheimers Research, Neurodegeneration, Neuroscience | |
| PBS (pH 7.3), 50% glycerol | |
| 1135 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Rat | |
| cholinergic receptor, nicotinic, alpha 2 (neuronal), cholinergic receptor, nicotinic, alpha polypeptide 2 (neuronal), neuronal acetylcholine receptor subunit alpha-2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 27-264 of human CHRNA2 (NP_000733.2). EEAKRPPPRAPGDPLSSPSPTALPQGGSHTETEDRLFKHLFRGYNRWARPVPNTSDVVIVRFGLSIAQLIDVDEKNQMMTTNVWLKQEWSDYKLRWNPTDFGNITSLRVPSEMIWIPDIVLYNNADGEFAVTHMTKAHLFSTGTVHWVPPAIYKSSCSIDVTFFPFDQQNCKMKFGSWTYDKAKIDLEQMEQTVDLKDYWESGEWAIVNATGTYNSKKYDCCAEIYPDVTYAFVIRRL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title