missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicotinic Acetylcholine R alpha 2/CHRNA2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94552-0.1ml
This item is not returnable.
View return policy
Description
Nicotinic Acetylcholine R alpha 2/CHRNA2 Polyclonal antibody specifically detects Nicotinic Acetylcholine R alpha 2/CHRNA2 in Rat samples. It is validated for Western Blot
Specifications
| Nicotinic Acetylcholine R alpha 2/CHRNA2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| cholinergic receptor, nicotinic, alpha 2 (neuronal), cholinergic receptor, nicotinic, alpha polypeptide 2 (neuronal), neuronal acetylcholine receptor subunit alpha-2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 27-264 of human CHRNA2 (NP_000733.2). EEAKRPPPRAPGDPLSSPSPTALPQGGSHTETEDRLFKHLFRGYNRWARPVPNTSDVVIVRFGLSIAQLIDVDEKNQMMTTNVWLKQEWSDYKLRWNPTDFGNITSLRVPSEMIWIPDIVLYNNADGEFAVTHMTKAHLFSTGTVHWVPPAIYKSSCSIDVTFFPFDQQNCKMKFGSWTYDKAKIDLEQMEQTVDLKDYWESGEWAIVNATGTYNSKKYDCCAEIYPDVTYAFVIRRL | |
| 0.1 mL | |
| Alzheimers Research, Neurodegeneration, Neuroscience | |
| 1135 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction