Learn More
Buerkle™ Nickle-Plated Brass Gastight Barrel Connector for Barrel Pump
Product Code. 10238230 Shop All Bürkle Products
Click to view available options
For Use With:
Stainless Steel Barrel Pump
Unit Size:
Each
Product Code. 10238230
Brand: Buerkle™ 56010130
Only null left
Add to basket
Add to basket
Buerkle™ Nickle-Plated Brass Gastight Barrel Connector for Barrel Pump prevents hazardous gases from escaping from the barrel.
  • The gas tight barrel screw joint prevents hazardous gases from escaping from the barrel
  • R 2 in. steel barrel thread, non-return valve with connection 1/8 in.
  • Nickel-plated brass, seals made of FKM
  • The container connectors make decanting even easier and safer with their more secure connection to barrels
  • The connectors secure the pump in the barrel, enable a vertical position change and allow the required air exchange
TRUSTED_SUSTAINABILITY
Specifications
Description Prevents hazardous gases from escaping
Includes Non-return valve with connection 1/8 in.
For Use With Stainless Steel Barrel Pump
Type Barrel Screw Joint
Material Nickel-plated Brass, FKM (Seals)