missing translation for 'onlineSavingsMsg'
Learn More

Nickel Nitrate GFAA Matrix Modifier: 1% Ni(NO3)2 in 2% HNO3, VHG™

Product Code. 18013030
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Product Code. Quantity unitSize
18013030 100 mL 100mL
1 options
This item is not returnable. View return policy

Product Code. 18013030

Brand: VHG Labs VHGMNIN1P100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Nickel Nitrate GFAA Matrix Modifier: 1% Ni(NO3)2 in 2% HNO3
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nickel Nitrate
CAS 13138-45-9
Methods AA/GFAA
Analyte or Component Names Nickel Nitrate
Shipping Condition Room Temperature
Molecular Formula Ni(NO3)2
Concentration 1%
Synonym Ni(NO3)2
SMILES [Ni+2].[O-][N+](=O)[O-].[O-][N+](=O)[O-]
IUPAC Name Nickel Nitrate
Solvent or Matrix Dilute HNO3
Molecular Weight (g/mol) 182.7
Quantity 100 mL
Formula Weight 182.7 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.