Learn More
Nickel(II) telluride, 99.9% (metals basis excluding Co), Co 0.1-1%, Thermo Scientific Chemicals
£66.60 - £230.00
Chemical Identifiers
CAS | 12142-88-0 |
---|---|
Molecular Formula | NiTe |
Molecular Weight (g/mol) | 186.29 |
MDL Number | MFCD00016258 |
InChI Key | NPEUSMKUOOTUGX-UHFFFAOYSA-N |
Synonym | nickel telluride, imgreite, ccris 7583, nickel telluride nickel and nickel compounds, nickeltellurid |
PubChem CID | 62780 |
IUPAC Name | tellanylidenenickel |
SMILES | [Ni]=[Te] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11302198
|
Thermo Scientific Alfa Aesar
013105.09 |
10 g |
£66.60
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11312198
|
Thermo Scientific Alfa Aesar
013105.18 |
50 g |
£230.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12142-88-0 | |
186.29 | |
NPEUSMKUOOTUGX-UHFFFAOYSA-N | |
62780 | |
[Ni]=[Te] |
NiTe | |
MFCD00016258 | |
nickel telluride, imgreite, ccris 7583, nickel telluride nickel and nickel compounds, nickeltellurid | |
tellanylidenenickel |
Specifications
12142-88-0 | |
NiTe | |
MFCD00016258 | |
nickel telluride, imgreite, ccris 7583, nickel telluride nickel and nickel compounds, nickeltellurid | |
[Ni]=[Te] | |
186.29 | |
186.31 | |
Odorless | |
Nickel(II) telluride |
Lump | |
NiTe | |
UN3077 | |
NPEUSMKUOOTUGX-UHFFFAOYSA-N | |
tellanylidenenickel | |
62780 | |
99.9% | |
(metals basis excluding Co), Co 0.1 to 1% |
Safety and Handling
GHS H Statement
H350-H372-H317
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H350i-H372
missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
missing translation for 'einecsNumber' : 235-260-6
missing translation for 'rtecsNumber' : QS0400000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only