missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) telluride, 99.9% (metals basis excluding Co), Co 0.1-1%

Product Code. 11312198
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11312198

Brand: Thermo Scientific Alfa Aesar 013105.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12142-88-0 | NiTe | 186.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12142-88-0
Molecular Formula NiTe
Molecular Weight (g/mol) 186.29
MDL Number MFCD00016258
InChI Key NPEUSMKUOOTUGX-UHFFFAOYSA-N
Synonym nickel telluride, imgreite, ccris 7583, nickel telluride nickel and nickel compounds, nickeltellurid
PubChem CID 62780
IUPAC Name tellanylidenenickel
SMILES [Ni]=[Te]

Specifications

Physical Form Lump
Quantity 50 g
Linear Formula NiTe
UN Number UN3077
Formula Weight 186.31
Percent Purity 99.9%
Odor Odorless
Assay (metals basis excluding Co), Co 0.1 to 1%
Chemical Name or Material Nickel(II) telluride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.