missing translation for 'onlineSavingsMsg'
Learn More

Nickel Electro™ Nickel Electro™ Nickel Crucible Lid

Nickel Crucible Lid

Brand:  Nickel Electro™ 53

Additional Details : Weight : 0.20000kg

Product Code. 16354972

  • £21.85 / Each
Estimated Shipment: 06-06-2024
to see stock.
Add to basket

Description

Description

  • Crucible lids, are flat with a depression
  • For centering on crucibles
  • Wwith an extension on one side that serves as a handle
  • Supplied in standard packs of 1
Specifications

Specifications

Nickel 201 Spec (Minimum 99.6% Nickel Content)
Autoclavable
Flat, circular with lip
Bright
80 mm
80 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers