missing translation for 'onlineSavingsMsg'
Learn More

Nickel Electro™ Nickel Electro™ Clifton™ Kosh Steam Steriliser

Product Code. 15445693 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
60 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric)
15445693 60 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15445693
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Electrically heated by a tubular immersion heater, completely manufactured from stainless steel. Uses steam to disinfect small, unwrapped instruments and other non-porous equipment not intended for clinical care of patients.

  • Featuring double skinned walled chamber for insulation purposes with two removable shelves for ease of loading
  • The lid is designed to ensure condensed steam runs back into the chamber
  • Lifting handles on both body and lid with an 18 mm diameter thermometer/thermocouple entry port
  • For stability mounted on four feet with filling/water level indicator reservoir for safety
  • The unit is protected by a thermal cut out which is manually reset
  • Drain Tap Fitted - Reservoir provides constant water level and drain point
  • Immersed Components - All stainless steel construction
  • Heater Power (220-240V) - 1000W
  • Power Consumption (220-240V) - 1000W
     

Specifications

Capacity (Metric) 60 L
Depth (Metric) Chamber 400 mm
Diameter (Metric) Chamber 400 mm
For Use With (Equipment) Melting Agar
Length (English) 17 in.
Height (English) Exterior 21 in.
Product Type Koch Steam Sterilizer
Weight (Metric) 24 kg
Size Large
Voltage 230 V
Sterility Non-sterile
Dimensions (L x W x H) 431 x 431 x 690 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.