missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Ni Sepharose™ High Performance Media

Product Code. 11535035 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11535035 25 mL 25mL
11545035 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11535035 Supplier Cytiva Supplier No. 17526801

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Precharged with Ni2+ for optimal purification of histidine-tagged proteins. Cytiva Lifescience™ Ni Sepharose™ High Performance Media shows high binding capacity and compatibility.

  • Negligible leakage of Ni2+ ions
  • High binding capacity: at least 40mg protein/mL medium
  • Compatible with a wide range of reducing agents, detergents, denaturants, and other additives
  • Stable (long-term) in the pH range 3-12
  • Average particle size: 34μm

TRUSTED_SUSTAINABILITY

Specifications

Product Type Ni Sepharose HP chromato graphy media
Quantity 25 mL
pH Range 3 to 12
Particle Size 34μm
Support Type 6% Cross-linked Agarose
Content And Storage 4° to 30°C
Matrix Ni-Sepharose
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • EUH208-Contains Nickel. May produce an allergic reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol, Nickel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.