missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Ni Sepharose™ 6 Fast Flow

Product Code. 11540814 Shop All Cytiva Products
Click to view available options
Quantity:
5 mL
25 mL
100 mL
500 mL
Unit Size:
100mL
25mL
500mL
5mL
This item is not returnable. View return policy

Product Code. 11540814

Brand: Cytiva 17531806

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Combines the advantages of Ni2 for purification of histidine tagged proteins, with the well industrially established properties of the Sepharose Fast Flow platform. Cytiva Lifescience™ Ni Sepharose™ 6 Fast Flow is part of a range of IMAC chromatography media for capture and intermediate purification.

Ni Sepharose 6 Fast Flow is a member of the Cytiva range of IMAC chromatography media for capture and intermediate purification.

  • Designed for scaling up purification of histidine-tagged proteins.
  • High protein binding capacity and minimal leakage of Ni2+ ions.
  • As a BioProcess medium it fulfills industrial demands for security of supply, robust performance and regulatory support.
  • Available in convenient prepacked HisPrep and HisTrap column formats.
  • Composed of cross-linked 6% agarose beads modified with a novel chelating ligand immobilized to the base matrix and pre-charged with Ni2+ ions for purification of histidine tagged proteins.
  • The coupling chemistry and the novel ligand is designed to give minimal ligand and Ni2+ leakage while providing high capacity.
  • Available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development.
  • As member of the BioProcess media range, Ni Sepharose 6 Fast Flow meets industrial demands with security of supply and comprehensive technical and regulatory support.

TRUSTED_SUSTAINABILITY

Specifications

Buffer 0.1M HCl, 0.1M NaOH, 8M Urea
Product Type Ni Sepharose 6 FF chromatography media
Quantity 5 mL
Height (English) 9.84 in.
Height (Metric) 250mm
Flow Rate 600 cm/hr.
Max. Pressure 1 bar
pH Range 3 to 12
Particle Size 90μm
Support Type 6% Cross-linked Agarose
Content And Storage 4° to 30°C
Matrix Ni-Sepharose
Show More Show Less
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.