missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NHERF-2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | NHERF-2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
NHERF-2 Polyclonal specifically detects NHERF-2 in Rat samples. It is validated for Western Blot.Specifications
| NHERF-2 | |
| Polyclonal | |
| Rabbit | |
| NP_446263 | |
| 9351 | |
| Synthetic peptide directed towards the N terminal of human Slc9a3r2. Peptide sequence RLVRGEQGYGFHLHGEKGRRGQFIRRVEPGSPAEAAALRAGDRLVEVNGV. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| E3KARP, Na(+)/H(+) exchange regulatory cofactor NHE-RF2, NHE3RF2, NHERF2MGC104639, NHERF-2NHE3 kinase A regulatory protein E3KARP, OCTS2, SIP-1SRY-interacting protein 1, sodium/hydrogen exchanger, Sodium-hydrogen exchanger regulatory factor 2, solute carrier family 9 (sodium/hydrogen exchanger), isoform 3 regulator 2, solute carrier family 9 (sodium/hydrogen exchanger), isoform 3 regulatoryfactor 2, solute carrier family 9 (sodium/hydrogen exchanger), member 3 regulator 2, Solute carrier family 9 isoform A3 regulatory factor 2, TKA-1SIP1, Tyrosine kinase activator protein 1 | |
| SLC9A3R2 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title