missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NHE8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18318-1-AP-150UL
This item is not returnable.
View return policy
Description
Sodium-hydrogen exchangers (NHEs), such as SLC9A8, are integral transmembrane proteins that exchange extracellular Na+ for intracellular H+. NHEs have multiple functions, including intracellular pH homeostasis, cell volume regulation, and electroneutral NaCl absorption in epithelia (Xu et al., 2008 ).Specifications
| NHE8 | |
| Polyclonal | |
| Unconjugated | |
| SLC9A8 | |
| KIAA0939, Na(+)/H(+) exchanger 8, NHE 8, NHE8, SLC9A8, Sodium/hydrogen exchanger 8 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23315, 311651, 77031 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q4L208, Q8R4D1, Q9Y2E8 | |
| Slc9a8 | |
| NHE8 Fusion Protein Ag13111 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction