missing translation for 'onlineSavingsMsg'
Learn More

NHE8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. p-7208748 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16878394 20 μL 20µL
16868394 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16878394 Supplier Proteintech Supplier No. 183181AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Sodium-hydrogen exchangers (NHEs), such as SLC9A8, are integral transmembrane proteins that exchange extracellular Na+ for intracellular H+. NHEs have multiple functions, including intracellular pH homeostasis, cell volume regulation, and electroneutral NaCl absorption in epithelia (Xu et al., 2008 ).

Specifications

Antigen NHE8
Applications Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SLC9A8
Gene Accession No. Q4L208, Q8R4D1, Q9Y2E8
Gene Alias KIAA0939, Na(+)/H(+) exchanger 8, NHE 8, NHE8, SLC9A8, Sodium/hydrogen exchanger 8
Gene Symbols Slc9a8
Host Species Rabbit
Immunogen NHE8 Fusion Protein Ag13111
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23315, 311651, 77031
Target Species Rat, Mouse, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.