Learn More
Invitrogen™ NFX1 Monoclonal Antibody (K16261_15F1), Invitrogen™
Mouse Monoclonal Antibody
£380.00 - £570.00
Specifications
| Antigen | NFX1 |
|---|---|
| Clone | K16261_15F1 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30209786
|
Invitrogen™
MA555832 |
50 μL |
£380.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30210267
|
Invitrogen™
MA556212 |
200 μL |
£570.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: FKFNTDAAEF IPQEKKNSGL NCGTQRRLDS NRIGRRNYSS PPPCHLSRQV PYDEISAVHQ HSYHPSGSKP KSQQTSFQSS PCNKSPKSHG LQNQPWQKLR NEKHHIRVKK AQSLAEQTSD TAGLESSTRS ESGTDLREHS PSESEKEVVG ADPRGAKPKK ATQFVYSYGR GPKVKGKLKC EWSNRTTPKP EDAGPESTKP VGVFHPDSSE ASSRKGVLDG YGARRNEQRR YPQKRPPWEV EGARPRPGRN PPKQEGHRHT NAGHRNNMGP IPKDDLNERP AKSTCDSENL AVINKSSRRV DQEKCTVRRQ DPQVVSPFSR GKQNHVLKNV ETHTGSLIEQ
NFX1 Monoclonal antibody specifically detects NFX1 in Human samples. It is validated for Western BlotSpecifications
| NFX1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 1300017N15Rik; 3000003M19Rik; DKFZp779G2416; MGC20369; m-Nfx.1; NFX.1; Nfx1; NFX2; nuclear transcription factor, X box-binding protein 1; nuclear transcription factor, X-box binding 1; TEG-42; testis expressed gene 42; Tex42; transcriptional repressor NF-X1; X-box binding 1 | |
| Recombinant human NFX1 (immunogen range is 11-350). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| NFX1 |
| K16261_15F1 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q12986 | |
| 4799 | |
| IgG2a | |
| Protein A | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.