Learn More
Invitrogen™ NFX1 Monoclonal Antibody (K16261_15F1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA555832
Description
Sequence of this protein is as follows: FKFNTDAAEF IPQEKKNSGL NCGTQRRLDS NRIGRRNYSS PPPCHLSRQV PYDEISAVHQ HSYHPSGSKP KSQQTSFQSS PCNKSPKSHG LQNQPWQKLR NEKHHIRVKK AQSLAEQTSD TAGLESSTRS ESGTDLREHS PSESEKEVVG ADPRGAKPKK ATQFVYSYGR GPKVKGKLKC EWSNRTTPKP EDAGPESTKP VGVFHPDSSE ASSRKGVLDG YGARRNEQRR YPQKRPPWEV EGARPRPGRN PPKQEGHRHT NAGHRNNMGP IPKDDLNERP AKSTCDSENL AVINKSSRRV DQEKCTVRRQ DPQVVSPFSR GKQNHVLKNV ETHTGSLIEQ
MHC class II gene expression is controlled primarily at the transcriptional level by transcription factors that bind to the X and Y boxes, two highly conserved elements in the proximal promoter of MHC class II genes. NFX1 is a transcriptional repressor capable of binding to the conserved X box motif of HLA-DRA and other MHC class II genes in vitro. The protein may play a role in regulating the duration of an inflammatory response by limiting the period in which class II MHC molecules are induced by IFN-gamma.MHC class II gene expression is controlled primarily at the transcriptional level by transcription factors that bind to the X and Y boxes, two highly conserved elements in the proximal promoter of MHC class II genes. The protein encoded by this gene is a transcriptional repressor capable of binding to the conserved X box motif of HLA-DRA and other MHC class II genes in vitro. The protein may play a role in regulating the duration of an inflammatory response by limiting the period in which class II MHC molecules are induced by IFN-gamma. Three alternative splice variants, each of which encodes a different isoform, have been identified.
Specifications
| NFX1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q12986 | |
| NFX1 | |
| Recombinant human NFX1 (immunogen range is 11-350). | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| K16261_15F1 | |
| Unconjugated | |
| NFX1 | |
| 1300017N15Rik; 3000003M19Rik; DKFZp779G2416; MGC20369; m-Nfx.1; NFX.1; Nfx1; NFX2; nuclear transcription factor, X box-binding protein 1; nuclear transcription factor, X-box binding 1; TEG-42; testis expressed gene 42; Tex42; transcriptional repressor NF-X1; X-box binding 1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 4799 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.