missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFkB1/NFkB p105 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
£175.00 - £391.00
Specifications
| Antigen | NFkB1/NFkB p105 |
|---|---|
| Concentration | 1.0 mg/ml |
| Dilution | Western Blot 1:1000, Simple Western 1:20, Immunohistochemistry 1:250, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:250, Immunohistochemistry-Frozen reported in scientific literature (PMID 26889045) |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18247865
|
Novus Biologicals
NBP1-77395SS |
0.025 mL |
£175.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18257775
|
Novus Biologicals
NBP1-77395 |
0.1 mL |
£391.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NFkB1/NFkB p105 Polyclonal specifically detects NFkB1/NFkB p105 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| NFkB1/NFkB p105 | |
| Western Blot 1:1000, Simple Western 1:20, Immunohistochemistry 1:250, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:250, Immunohistochemistry-Frozen reported in scientific literature (PMID 26889045) | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse | |
| P19838, P25799 | |
| 4790 | |
| A genomic peptide made to an N-terminal region of the human NFkB p105/p50 protein (within residues 400-590). [Swiss-Prot P19838] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1.0 mg/ml | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| DNA-binding factor KBF1, EBP-1, NF-kappa-B, NF-kB1, NFKB1, NFKB-p105, Nuclear factor NF-kappa-B p105 subunit, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 1, p105 | |
| NFKB1 | |
| IgG | |
| Affinity Purified | |
| 105 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title