missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFkB1/NFkB p105 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
Brand: Novus Biologicals NBP1-77395SS
This item is not returnable.
View return policy
Description
NFkB1/NFkB p105 Polyclonal specifically detects NFkB1/NFkB p105 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| NFkB1/NFkB p105 | |
| Polyclonal | |
| Unconjugated | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| DNA-binding factor KBF1, EBP-1, NF-kappa-B, NF-kB1, NFKB1, NFKB-p105, Nuclear factor NF-kappa-B p105 subunit, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 1, p105 | |
| Rabbit | |
| 105 kDa | |
| 0.025 mL | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/ml | |
| Western Blot 1:1000, Simple Western 1:20, Immunohistochemistry 1:250, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:250, Immunohistochemistry-Frozen reported in scientific literature (PMID 26889045) | |
| P19838, P25799 | |
| NFKB1 | |
| A genomic peptide made to an N-terminal region of the human NFkB p105/p50 protein (within residues 400-590). [Swiss-Prot P19838] | |
| Affinity Purified | |
| RUO | |
| 4790 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction