missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFIB Rabbit anti-Human, Mouse, Clone: 7T1G5, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | NFIB |
|---|---|
| Clone | 7T1G5 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18372096
|
Novus Biologicals
NBP3-16757-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18315083
|
Bio-Techne
NBP3-16757-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NFIB Monoclonal antibody specifically detects NFIB in Human, Mouse samples. It is validated for Western BlotSpecifications
| NFIB | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 4781 | |
| IgG | |
| Affinity purified |
| 7T1G5 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| CCAAT-box-binding transcription factor, CTF, HMGIC/NFIB, NF-I/B, NFI-B, NFIB2, NFIB3, NFI-RED, nuclear factor 1 B-type, Nuclear factor 1/B, nuclear factor I/BNF1-B, TGGCA-binding protein | |
| A synthetic peptide corresponding to a sequence within amino acids 321-420 of human NFIB (O00712). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title