missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFIB Rabbit anti-Human, Mouse, Clone: 7T1G5, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Bio-Techne NBP3-16757-100UL
This item is not returnable.
View return policy
Description
NFIB Monoclonal antibody specifically detects NFIB in Human, Mouse samples. It is validated for Western Blot
Specifications
| NFIB | |
| Monoclonal | |
| Unconjugated | |
| CCAAT-box-binding transcription factor, CTF, HMGIC/NFIB, NF-I/B, NFI-B, NFIB2, NFIB3, NFI-RED, nuclear factor 1 B-type, Nuclear factor 1/B, nuclear factor I/BNF1-B, TGGCA-binding protein | |
| A synthetic peptide corresponding to a sequence within amino acids 321-420 of human NFIB (O00712). | |
| 100 μg | |
| Cell Cycle and Replication | |
| 4781 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 7T1G5 | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction