missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NFE2 Recombinant Rabbit Monoclonal Antibody (24GB520)
Rabbit Recombinant Monoclonal Antibody
£126.00 - £434.00
Specifications
| Antigen | NFE2 |
|---|---|
| Clone | 24GB520 |
| Concentration | 0.6 mg/mL |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30275720
|
Invitrogen™
MA557115 |
100 μL |
£434.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30276644
|
Invitrogen™
MA557116 |
20 μL |
£126.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NFE2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Component of the NF-E2 complex essential for regulating erythroid and megakaryocytic maturation and differentiation. Binds to the hypersensitive site 2 (HS2) of the beta-globin control region (LCR). This subunit (NFE2) recognizes the TCAT/C sequence of the AP-1-like core palindrome present in a number of erythroid and megakaryocytic gene promoters. Requires MAFK or other small MAF proteins for binding to the NF-E2 motif. May play a role in all aspects of hemoglobin production from globin and heme synthesis to procurement of iron.Specifications
| NFE2 | |
| 0.6 mg/mL | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q07279, Q16621, Q6AYT2 | |
| 18022, 366998, 4778 | |
| A synthesized peptide derived from human NFE2 (aa 275-334). | |
| Antibody |
| 24GB520 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| NFE2 | |
| leucine zipper protein NF-E2; NFE2; NF-E2; NF-E2/P45; nuclear factor (erythroid-derived 2), 45kD; nuclear factor (erythroid-derived 2), 45kDa; nuclear factor, erythroid 2; nuclear factor, erythroid derived 2; nuclear factor, erythroid-derived 2 45 kDa subunit; p45; p45 NF-E2; p45NFE2; p45nf-e2; Transcription factor NF-E2 45 kDa subunit | |
| NFE2 | |
| Primary | |
| Affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title