Learn More
Invitrogen™ NFE2 Recombinant Rabbit Monoclonal Antibody (24GB520)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557115
 View more versions of this product
Product Code. 30275720
  • £434.00 / 100µL
Description
NFE2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Component of the NF-E2 complex essential for regulating erythroid and megakaryocytic maturation and differentiation. Binds to the hypersensitive site 2 (HS2) of the beta-globin control region (LCR). This subunit (NFE2) recognizes the TCAT/C sequence of the AP-1-like core palindrome present in a number of erythroid and megakaryocytic gene promoters. Requires MAFK or other small MAF proteins for binding to the NF-E2 motif. May play a role in all aspects of hemoglobin production from globin and heme synthesis to procurement of iron.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NFE2
Recombinant Monoclonal
0.6 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q07279, Q16621, Q6AYT2
NFE2
A synthesized peptide derived from human NFE2 (aa 275-334).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB520
Unconjugated
NFE2
leucine zipper protein NF-E2; NFE2; NF-E2; NF-E2/P45; nuclear factor (erythroid-derived 2), 45kD; nuclear factor (erythroid-derived 2), 45kDa; nuclear factor, erythroid 2; nuclear factor, erythroid derived 2; nuclear factor, erythroid-derived 2 45 kDa subunit; p45; p45 NF-E2; p45NFE2; p45nf-e2; Transcription factor NF-E2 45 kDa subunit
Rabbit
Affinity chromatography
RUO
18022, 366998, 4778
-20°C