missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFE2 Mouse anti-Human, Clone: 1D7C8, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | NFE2 |
|---|---|
| Clone | 1D7C8 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16818315
|
Proteintech
66436-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16808315
|
Proteintech
66436-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Component of the NF-E2 complex essential for regulating erythroid and megakaryocytic maturation and differentiation. Binds to the hypersensitive site 2 (HS2) of the beta-globin control region (LCR). This subunit (NFE2) recognizes the TCAT/C sequence of the AP-1-like core palindrome present in a number of erythroid and megakaryocytic gene promoters. Requires MAFK or other small MAF proteins for binding to the NF-E2 motif. May play a role in all aspects of hemoglobin production from globin and heme synthesis to procurement of iron.Specifications
| NFE2 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Leucine zipper protein NF E2, NF E2, NFE2, p45, p45 NF E2 | |
| NFE2 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 1D7C8 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q16621 | |
| 4778 | |
| NFE2 Fusion Protein Ag1570 | |
| Primary | |
| -20°C | |
| NFE2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title