missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFATC1 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | NFATC1 |
|---|---|
| Concentration | 650 μg/mL |
| Applications | Western Blot, Immunofluorescence, ELISA |
| Form | Liquid |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18087305
|
Proteintech
30114-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18020356
|
Proteintech
30114-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The NFATC1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human NFATC1. This antibody recognizes human antigen. The NFATC1 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
| NFATC1 | |
| Western Blot, Immunofluorescence, ELISA | |
| Rabbit | |
| 4772 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
| 650 μg/mL | |
| Liquid | |
| Human | |
| NF ATC, NF ATc1, NFAT2, NFATc, NFATC1 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title