Learn More
NFATC1 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30114-1-AP-150UL
This item is not returnable.
View return policy
Description
The NFATC1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human NFATC1. This antibody recognizes human antigen. The NFATC1 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
| NFATC1 | |
| 650 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, Immunofluorescence, ELISA | |
| NF ATC, NF ATc1, NFAT2, NFATc, NFATC1 | |
| 150 μL | |
| 4772 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |