missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NFAT5 Monoclonal Antibody (K49018_1C5), Invitrogen™

Mouse Monoclonal Antibody

£380.00 - £570.00

Specifications

Antigen NFAT5
Clone K49018_1C5
Concentration 1 mg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30205379
View Documents
Invitrogen™
MA556149
200 μL
£570.00
200µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30213099
View Documents
Invitrogen™
MA555769
50 μL
£380.00
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sequence of this protein is as follows: GQYPVKSEGK ELKIVVQPET QHRARYLTEG SRGSVKDRTQ QGFPTVKLEG HNEPVVLQVF VGNDSGRVKP HGFYQACRVT GRNTTPCKEV DIEGTTVIEV GLDPSNNMTL AVDCVGILKL RNADVEARIG IAGSKKKSTR ARLVFRVNIM RKDGSTLTLQ TPSSPILCTQ PAGVPEILKK SLHSCSVKGE EEVFLIGKNF LKGTKVIFQE NVSDENSWKS EAEIDMELFH QNHLIVKVPP YHDQHITLPV SVGIYVVTNA GRSHDVQPFT

NFAT5 Monoclonal antibody specifically detects NFAT5 in Human samples. It is validated for Western Blot
Specifications

Specifications

NFAT5
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
AI225870; B130038B15Rik; CAG-8; CAG80; glutamine rich protein H65; KIAA0827; mKIAA0827; Nfat; Nfat5; NF-AT5; NFATL1; NFAT-like protein 1; nfatz; nuclear factor of activated T cells 5; nuclear factor of activated T-cells 5; nuclear factor of activated T-cells 5, tonicity-responsive; OREBP; osmotic response element-binding protein; rel domain-containing transcription factor NFAT5; T-cell transcription factor NFAT5; TonE-binding protein; TONEBP; tonicity-responsive enhancer binding protein; tonicity-responsive enhancer-binding protein
Recombinant human NFAT5 (immunogen range is 271-540).
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
NFAT5
K49018_1C5
Western Blot
Unconjugated
Mouse
Human
O94916
10725
IgG2a
Protein A
Antibody
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.