missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NFAT5 Monoclonal Antibody (K49018_1C5), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA556149

 View more versions of this product

Product Code. 30205379

  • £586.00 / 200µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence of this protein is as follows: GQYPVKSEGK ELKIVVQPET QHRARYLTEG SRGSVKDRTQ QGFPTVKLEG HNEPVVLQVF VGNDSGRVKP HGFYQACRVT GRNTTPCKEV DIEGTTVIEV GLDPSNNMTL AVDCVGILKL RNADVEARIG IAGSKKKSTR ARLVFRVNIM RKDGSTLTLQ TPSSPILCTQ PAGVPEILKK SLHSCSVKGE EEVFLIGKNF LKGTKVIFQE NVSDENSWKS EAEIDMELFH QNHLIVKVPP YHDQHITLPV SVGIYVVTNA GRSHDVQPFT

The nuclear factor of activated T-cells (NFAT) transcription complex is required for the expression of a group of proteins that collectively regulate the immune response. Four NFAT proteins, encoded on separate genes and expressed as several splice variants, have been described: NFAT1 (also known as NFATp or NFATc2), NFAT2 (NFATc or NFATc1), NFAT3, and NFAT4 (NFATx or NFATc3). These proteins show a low level of sequence similarity with the Dorsal/Rel/NFkB family of transcription factors. Another NFAT-related protein termed NFAT5 differs from isoforms 1-4 in that it lacks many of the Fos/Jun contact sites observed in its predecessors and its subcellular localization is not calcineurin-dependent.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NFAT5
Monoclonal
1 mg/mL
PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
O94916
NFAT5
Recombinant human NFAT5 (immunogen range is 271-540).
200 μL
Primary
Human
Antibody
IgG2a
Western Blot
K49018_1C5
Unconjugated
NFAT5
AI225870; B130038B15Rik; CAG-8; CAG80; glutamine rich protein H65; KIAA0827; mKIAA0827; Nfat; Nfat5; NF-AT5; NFATL1; NFAT-like protein 1; nfatz; nuclear factor of activated T cells 5; nuclear factor of activated T-cells 5; nuclear factor of activated T-cells 5, tonicity-responsive; OREBP; osmotic response element-binding protein; rel domain-containing transcription factor NFAT5; T-cell transcription factor NFAT5; TonE-binding protein; TONEBP; tonicity-responsive enhancer binding protein; tonicity-responsive enhancer-binding protein
Mouse
Protein A
RUO
10725
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.