missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NF-kB Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14220-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody can bind both p105 and p50 isoforms of NFKB1.
NF-kB is a nuclear transcription factor activated by various extra and intracellular stimuli such as cytokines, UV radiation, stress, in injury, and by bacterial and viral products. It is involved in regulation of various cellular events including cell growth, differentiation, proliferation, apoptosis and inflammation. NFKB1 (p50), a 50 kDa functional sub-unit of NF-kB, is a member of Rel protein family. It is synthesized as a p105 precursor protein and consists of an N-terminal conserved RHD-region containing nuclear localization signal, DNA-binding and dimerization domains. NFKB1 (p50) forms homodimers or heterodimerizes with p65, forming the functional NF-kB factor. NFKB1 (p50) directs the nuclear translocation of NF-kB and is instrumental in its DNA-binding. Pathological role of NF-kB has been suggested in AIDS, hematogenic cancer cell metastasis and rheumatoid arthritis.Specifications
| NF-kB | |
| Polyclonal | |
| Unconjugated | |
| NFKB1 | |
| DNA binding factor KBF1, EBP 1, KBF1, NF kappa B, NFKB p105, NFKB p50, NFKB1, NFKB1,p105, NFKB1, p105, p50, nfkb1a, NF-kappaB, NF-kappaB 1, p105, p50 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4790, 81736 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Western Blot, Immunoprecipitation, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P19838, Q63369 | |
| NFKB1 | |
| NF-kappaB Fusion Protein Ag5458 | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction