missing translation for 'onlineSavingsMsg'
Learn More

CellSensor™ CellSensor™ NFκB-bla THP-1 Cell Line

Product Code. 10266743
Click to view available options
Quantity:
1 vial
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10266743

Brand: CellSensor™ K1662

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CellSensor™ NFκB-bla THP-1 Cell Line contains a beta-lactamase reporter gene under the control of the Nuclear Factor kappa B response element.

  • The CellSensor™ NFκB-bla THP-1 cell line contains a beta-lactamase reporter gene under the control of the Nuclear Factor kappa B (NFκB) response element stably integrated into THP-1 cells.
  • To obtain the cell line, the NFκB-bla construct was transduced into THP-1 cells by lentivirus.
  • Subsequent flow cytometry was used to isolate cells responsive to Tumor Necrosis Factor alpha (TNFα).
  • The cell line was validated for DMSO tolerance, cell number, stimulation time, substrate loading time, and EC50 concentration of TNFα.
  • The CellSensor™ NFκB-bla THP-1 cell line is responsive to TNFα and can be used to probe NFκB signaling pathways, including those involved in apoptosis, viral defense, cancer, inflammation, and autoimmune disease.
  • This cell line can be adapted for high-throughput screening for agonists or antagonists with compound libraries.
  • Candidate drugs can be tested for dose response against this cell line.
TRUSTED_SUSTAINABILITY

Specifications

Assay Entry Cell-based beta lactamase reporter gene
Cell State Dividing Cells
Content And Storage CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.
Product Line CellSensor
Quantity 1 vial
Target Entry TNF alpha, NIK, TLR, I kappa B, IRAK, IKK, NF-kappa B pathway
Type Cell Line
Cell Line THP-1
System Type CellSensor™
Druggable Target Signaling Pathway

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.