Learn More
NeutraKine™ PDGF-BB Mouse anti-Human, Unconjugated, Clone: 4G11C7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 69020-1-IG-25UG
Description
The NeutraKine™ PDGF-BB antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human NeutraKine™ PDGF-BB. This antibody recognizes human antigen. The NeutraKine™ PDGF-BB antibody has been validated for the following applications: Neutralization, ELISA analysis.
Specifications
| NeutraKine™ PDGF-BB | |
| Monoclonal | |
| Unconjugated | |
| 25 μg | |
| 5155 | |
| IgG1 |
| Neutralization | |
| 4G11C7 | |
| Mouse | |
| Primary | |
| Human |