Learn More
NeutraKine™ PDGF-BB Mouse anti-Human, Unconjugated, Clone: 4G11C7, Proteintech
Mouse Monoclonal Antibody
£99.00 - £249.00
Specifications
| Antigen | NeutraKine™ PDGF-BB |
|---|---|
| Clone | 4G11C7 |
| Applications | Neutralization |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17521496
|
Proteintech
69020-1-IG-25UG |
25 μg |
£99.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17511496
|
Proteintech
69020-1-IG-100UG |
100 μg |
£249.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The NeutraKine™ PDGF-BB antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human NeutraKine™ PDGF-BB. This antibody recognizes human antigen. The NeutraKine™ PDGF-BB antibody has been validated for the following applications: Neutralization, ELISA analysis.
Specifications
| NeutraKine™ PDGF-BB | |
| Neutralization | |
| Unconjugated | |
| Human | |
| IgG1 |
| 4G11C7 | |
| Monoclonal | |
| Mouse | |
| 5155 | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.