missing translation for 'onlineSavingsMsg'
Learn More

PeproTech Neurturin Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
GREENER_CHOICE

Rabbit Polyclonal Antibody

£177.00 - £2745.00

Specifications

Antigen Neurturin
Concentration 0.1-1.0 mg/mL
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Biotin
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17884793
GREENER_CHOICE
View Documents
PeproTech
500-P102BT-25UG
25 μg
£177.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17893853
View Documents
PeproTech
500-P102BT-50UG
50 μg
£228.00
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17874783
View Documents
Invitrogen
500-P102BT-1MG
2 x 500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
30054493
View Documents
PeproTech
500-P102BT-500UG
500 μg
£1466.00
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

AA Sequence of recombinant protein: ARLGARPCGL RELEVRVSEL GLGYASDETV LFRYCAGACE AAARVYDLGL RRLRQRRRLR RERVRAQPCC RPTAYEDEVS FLDAHSRYHT VHELSARECA CV. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human Neurturin. Anti-Human Neurturin-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human Neurturin by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human Neurturin (500-P102) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human Neurturin. Western Blot: To detect hNeurturin by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hNeurturin is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P102BT-1MG will be provided as 2 x 500 μg

Glial cell line-derived neurotrophic factor (GDNF) plays key roles in the control of vertebrate neuron survival and differentiation. A novel neurotrophic factor was recently cloned from human and mouse and designated neurturin. Physiological responses to neurturin (NTN) require the presence of receptor tyrosine kinase RET and a novel glycosylphosphatidylinositol linked receptor NTNRalpha. The cDNAs encoding NTNRalpha from human, rat, chicken, and mouse have been cloned recently and termed GDNFR-b, Ret ligand 2 (RETL2) or TGF-beta-related neurotrophic factor receptor 2 (TrnR2) and nominated as GFR-a-2 recently. NTN binds to and forms a complex with GFR-a-2 and the Ret PTK and activates the RET receptor tyrosine kinase pathway. Both NTN and GDNF can activate the MAP kinase and phosphatidylinositol 3-kinase pathways and play a critical role in the development of many neuronal populations. Neurturin and GDNF define a new family of neurotrophic factors.
Specifications

Specifications

Neurturin
ELISA, Western Blot
Biotin
Rabbit
Human
Q99748
4902
E.coli-derived, 23.6 kDa Recombinant Human Neurturin.
Antigen affinity chromatography
NRTN
0.1-1.0 mg/mL
Polyclonal
Lyophilized
RUO
PBS with no preservative
neurturin; NRTN; NTN; prepro-neurturin
NRTN
Primary
-20°C
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.