Learn More
PeproTech Neurturin Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody
Brand: PeproTech 500-P102BT-25UG
Description
AA Sequence of recombinant protein: ARLGARPCGL RELEVRVSEL GLGYASDETV LFRYCAGACE AAARVYDLGL RRLRQRRRLR RERVRAQPCC RPTAYEDEVS FLDAHSRYHT VHELSARECA CV. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human Neurturin. Anti-Human Neurturin-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human Neurturin by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human Neurturin (500-P102) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human Neurturin. Western Blot: To detect hNeurturin by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hNeurturin is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P102BT-1MG will be provided as 2 x 500 μg
Glial cell line-derived neurotrophic factor (GDNF) plays key roles in the control of vertebrate neuron survival and differentiation. A novel neurotrophic factor was recently cloned from human and mouse and designated neurturin. Physiological responses to neurturin (NTN) require the presence of receptor tyrosine kinase RET and a novel glycosylphosphatidylinositol linked receptor NTNRalpha. The cDNAs encoding NTNRalpha from human, rat, chicken, and mouse have been cloned recently and termed GDNFR-b, Ret ligand 2 (RETL2) or TGF-beta-related neurotrophic factor receptor 2 (TrnR2) and nominated as GFR-a-2 recently. NTN binds to and forms a complex with GFR-a-2 and the Ret PTK and activates the RET receptor tyrosine kinase pathway. Both NTN and GDNF can activate the MAP kinase and phosphatidylinositol 3-kinase pathways and play a critical role in the development of many neuronal populations. Neurturin and GDNF define a new family of neurotrophic factors.
Specifications
| Neurturin | |
| Polyclonal | |
| Biotin | |
| NRTN | |
| neurturin; NRTN; NTN; prepro-neurturin | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4902 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| Q99748 | |
| NRTN | |
| E.coli-derived, 23.6 kDa Recombinant Human Neurturin. | |
| 25 μg | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.