Learn More
R&D Systems™ NeuroTACS In Situ Apoptosis Detection Kit
Description
Includes
Proteinase K, 30uL; NeuroPore™, 5mL; TdT Enzyme, 30uL; 50X Mn+2, 30uL; Blue counterstain, 50mL; TACS-Nuclease and buffer; dNTP mix and buffers for TdT, DAB solution and enhancer
Primary neuronal cell cultures are often maintained in mixed culture, therefore, the ability to identify specific cells using criterion other than morphology is important. Similarly, it can be difficult to identify cell types within central nervous system
Complete kits contain Streptavidin-HRP, DAB, Blue Counterstain, and reagents necessary for generating positive controls
Study fixed cells, embedded tissues and frozen tissues using light microscopy
Specifications
Specifications
| Quantity | 30 Tests |
| Content And Storage | The product requires storage at -20 to -70° C, 2 - 8° C, and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date. |