missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neurofibromin 1 Antibody (McNFn27a) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
£175.00 - £382.00
Specifications
| Antigen | Neurofibromin 1 |
|---|---|
| Clone | McNFn27a |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 2-5 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry 1:20 - 1:50, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:20 - 1:50. Use reported in scientific literature |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18224810
|
Novus Biologicals
NB300-153SS |
0.05 mL |
£175.00
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18060964
|
Novus Biologicals
NB300-153 |
0.2 mL |
£382.00
0.20mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Neurofibromin 1 Monoclonal specifically detects Neurofibromin 1 in Human, Mouse, Rat, Guinea Pig samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| Neurofibromin 1 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Guinea Pig | |
| DKFZp686J1293, FLJ21220, Neurofibromatosis-related protein NF-1, neurofibromin, neurofibromin 1, NFNS, VRNF, WSS | |
| NF1 | |
| IgG1 κ | |
| Affinity Purified | |
| This is specific for full-length or truncated forms of neurofibromin. |
| McNFn27a | |
| Western Blot 2-5 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry 1:20 - 1:50, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:20 - 1:50. Use reported in scientific literature | |
| Monoclonal | |
| Mouse | |
| Cancer, Cell Cycle and Replication, Chromatin Research, Cytoskeleton Markers, Hypoxia, Neuroscience, Neurotransmission, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors | |
| P21359 | |
| 4763 | |
| A highly conserved N-terminal peptide, corresponding to amino acids 27-41 of human Neurofibromin 1. [UniProt# P21359] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title