missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neurofibromin 1 Antibody (McNFn27a) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB300-153
This item is not returnable.
View return policy
Description
Neurofibromin 1 Monoclonal specifically detects Neurofibromin 1 in Human, Mouse, Rat, Guinea Pig samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| Neurofibromin 1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 2-5 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry 1:20 - 1:50, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:20 - 1:50. Use reported in scientific literature | |
| DKFZp686J1293, FLJ21220, Neurofibromatosis-related protein NF-1, neurofibromin, neurofibromin 1, NFNS, VRNF, WSS | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This is specific for full-length or truncated forms of neurofibromin. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| McNFn27a | |
| Unconjugated | |
| P21359 | |
| NF1 | |
| A highly conserved N-terminal peptide, corresponding to amino acids 27-41 of human Neurofibromin 1. [UniProt# P21359] | |
| 0.2 mL | |
| Cancer, Cell Cycle and Replication, Chromatin Research, Cytoskeleton Markers, Hypoxia, Neuroscience, Neurotransmission, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors | |
| 4763 | |
| Human, Mouse, Rat, Guinea Pig | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction