missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neuroblastoma Antibody (FMG25) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50528-0.025ml
This item is not returnable.
View return policy
Description
Neuroblastoma Monoclonal antibody specifically detects Neuroblastoma in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| Neuroblastoma | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry 1:10 - 1:1000, CyTOF-ready | |
| ARHGEF16, EPHEXIN4, GEF16, NBR | |
| Human cutaneous T cell lymphoma (HUT 78), SPF female Balb/c mice were injected i.p with 2 x 10*5 cells in complete Freund's adjuvant. 30 days later a 2nd i.p injection of 2 x 10*5 cells without adjuvant was given. Fusion was 5 days | |
| 0.025 mL | |
| Cancer | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| FMG25 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction