missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neuroblastoma Antibody (FMG25) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£150.00 - £302.00
Specifications
| Antigen | Neuroblastoma |
|---|---|
| Clone | FMG25 |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry 1:10 - 1:1000, CyTOF-ready |
| Applications | Flow Cytometry, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18654399
|
Novus Biologicals
NBP2-50528-0.025ml |
0.025 mL |
£150.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18673869
|
Novus Biologicals
NBP2-50528 |
0.1 mL |
£302.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Neuroblastoma Monoclonal antibody specifically detects Neuroblastoma in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
| Neuroblastoma | |
| 1 mg/mL | |
| Flow Cytometry, CyTOF | |
| Unconjugated | |
| Mouse | |
| Cancer | |
| PBS | |
| Human cutaneous T cell lymphoma (HUT 78), SPF female Balb/c mice were injected i.p with 2 x 10*5 cells in complete Freund's adjuvant. 30 days later a 2nd i.p injection of 2 x 10*5 cells without adjuvant was given. Fusion was 5 days | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| FMG25 | |
| Flow Cytometry 1:10 - 1:1000, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ARHGEF16, EPHEXIN4, GEF16, NBR | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title