missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neuro D4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-80040
This item is not returnable.
View return policy
Description
Neuro D4 Polyclonal specifically detects Neuro D4 in Human, Rat samples. It is validated for Western Blot.
Specifications
| Neuro D4 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| BAF45B, BRG1-associated factor 45B, D4, zinc and double PHD fingers family 1NEUD4BAF45b, MGC150428, MGC150429, neuro-d4, neuro-d4 homolog, zinc finger protein neuro-d4 | |
| Rabbit | |
| 40 kDa | |
| 100 μL | |
| Apoptosis | |
| 8193 | |
| Human, Rat, Rat, Bovine, Canine, Equine, Guinea Pig | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_004638 | |
| DPF1 | |
| Synthetic peptide directed towards the middle region of human DPF1. Peptide sequence KELAWVPEAQRKHTAKKAPDGTVIPNGYCDFCLGGSKKTGCPEDLISCAD. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction