Learn More
Invitrogen™ Neural/Glial Antigen 2 (NG2) Monoclonal Antibody (9.2.27), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£1376.00 - £3115.00
Specifications
| Antigen | Neural/Glial Antigen 2 (NG2) |
|---|---|
| Clone | 9.2.27 |
| Concentration | 0.5 mg/mL |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
This 9.2.27 MAb reacts with human neural/glial antigen 2, which is also known as melanoma chondroitin sulfate proteoglycan (MCSP). This antigen is composed of a 250 kDa N-linked glycoprotein and a >450 kDa proteoglycan. NG2 is present on the surface of >90% of malignant melanomas in addition to some non-melanomic tumors. Moreover, NG2 is expressed on glioma cells, as well as on developing and adult oligodendrocyte precursor cells. Studies have demonstrated the involvement of this protein in tumor cell proliferation, adhesion, migration, and invasion. NG2 also functions as a coreceptor for alpha4beta1 integrin. Finally, expression of NG2 can be used as a prognostic marker for disorders such as acral letiginous melanoma and infantile acute myeloid leukemia. The 9.2.27 antibody has been reported to suppress melanoma tumor growth.
A human melanoma-associated chondroitin sulfate proteoglycan plays a role in stabilizing cell-substratum interactions during early events of melanoma cell spreading on endothelial basement membranes. CSPG4 represents an integral membrane chondroitin sulfate proteoglycan expressed by human malignant melanoma cells.Specifications
| Neural/Glial Antigen 2 (NG2) | |
| 0.5 mg/mL | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q6UVK1 | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
| 9.2.27 | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| 1464 | |
| Primary | |
| 4°C | |
| Cspg4 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.