missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Netrin-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £386.00
Specifications
| Antigen | Netrin-1 |
|---|---|
| Dilution | Western Blot : 5-2 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30067044
|
Novus Biologicals™
NBP3-30884-100ug |
100 μg |
£386.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30067033
|
Novus Biologicals™
NBP3-30884-25ug |
25 μg |
£162.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Netrin-1 Polyclonal antibody specifically detects Netrin-1 in Bovine,Feline,Human,Mouse,Rat,Macaque samples. It is validated for Western BlotSpecifications
| Netrin-1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis | |
| Phosphate buffered saline | |
| 9423 | |
| IgG | |
| Ammonium sulfate precipitation |
| Western Blot : 5-2 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Bovine, Feline, Human, Mouse, Rat, Macaque | |
| netrin 1, netrin-1, NTN1Lnetrin 1, mouse, homolog of | |
| Synthetic peptide derived from the human Netrin-1 | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title