missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Netrin-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30884-25ug
This item is not returnable.
View return policy
Description
Netrin-1 Polyclonal antibody specifically detects Netrin-1 in Bovine,Feline,Human,Mouse,Rat,Macaque samples. It is validated for Western Blot
Specifications
| Netrin-1 | |
| Polyclonal | |
| Western Blot : 5-2 μg/mL | |
| netrin 1, netrin-1, NTN1Lnetrin 1, mouse, homolog of | |
| Synthetic peptide derived from the human Netrin-1 | |
| 25 μg | |
| Apoptosis | |
| 9423 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Bovine, Feline, Human, Mouse, Rat, Macaque | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction