missing translation for 'onlineSavingsMsg'
Learn More

Netilmicin Sulfate, TRC

Product Code. 18080920
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18080920 10 mg 10mg
18043530 25 mg 25mg
18062030 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18080920 Supplier Toronto Research Chemicals Supplier No. TRCN39080010MG
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56391-57-2
Molecular Formula 2 C21 H41 N5 O7 . 5 H2 O4 S
Molecular Weight (g/mol) 1441.55
Synonym Netilmicin sulfate, 2-Deoxy-6-O-[3-deoxy-4-C-methyl-3-(methylamino)-β-l-arabinopyranosyl]-4-O-(2,6-diamino-2,3,4,6-tetradeoxy-α-d-glycero-hex-4-enopyranosyl)-1-N-ethyl-d-streptamine sulfate, 2-Deoxy-6-O-[3-deoxy-4-C-methyl-3-(methylamino)-beta-l-arabinopyranosyl]-4-O-(2,6-diamino-2,3,4,6-tetradeoxy-alpha-d-glycero-hex-4-enopyranosyl)-1-N-ethyl-d-streptamine sulfate
IUPAC Name (2R,3R,4R,5R)-2-[(1S,2S,3R,4S,6R)-4-amino-3-[[(2S,3R)-3-amino-6-(aminomethyl)-3,4-dihydro-2H-pyran-2-yl]oxy]-6-(ethylamino)-2-hydroxycyclohexyl]oxy-5-methyl-4-(methylamino)oxane-3,5-diol;sulfuric acid
SMILES CCN[C@@H]1C[C@H](N)[C@@H](O[C@H]2OC(=CC[C@H]2N)CN)[C@H](O)[C@H]1O[C@H]3OC[C@](C)(O)[C@H](NC)[C@H]3O.CCN[C@@H]4C[C@H](N)[C@@H](O[C@H]5OC(=CC[C@H]5N)CN)[C@H](O)[C@H]4O[C@H]6OC[C@](C)(O)[C@H](NC)[C@H]6O.OS(=O)(=O)O.OS(=O)(=O)O.OS(=O)(=O)O.OS(=O)(=O)O.OS(=O)(=O)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/2C21H41N5O7.5H2O4S/c2*1-4-26-13-7-12(24)16(32-19-11(23)6-5-10(8-22)31-19)14(27)17(13)33-20-15(28)18(25-3)21(2,29)9-30-20;5*1-5(2,3)4/h2*5,11-20,25-29H,4,6-9,22-24H2,1-3H3;5*(H2,1,2,3,4)/t2*11-,12+,13-,14+,15-,16-,17+,18-,19-,20-,21+;;;;;/m11...../s1
Formula Weight 1440.4381
Chemical Name or Material Netilmicin sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.