accessibility menu, dialog, popup

UserName

CAS RN 56391-57-2

IUPAC Name: bis((2R,3R,4R,5R)-2-{[(1S,2S,3R,4S,6R)-4-amino-3-{[(2S,3R)-3-amino-6-(aminomethyl)-3,4-dihydro-2H-pyran-2-yl]oxy}-6-(ethylamino)-2-hydroxycyclohexyl]oxy}-5-methyl-4-(methylamino)oxane-3,5-diol); pentakis(sulfuric acid)
Synonyms: netilmicin
Molecular Weight (g/mol): 1441.53
Molecular Formula: C42H92N10O34S5
InChi Key: AGFWIZQEWFGATK-UNZHCMSXSA-N
SMILES: OS(O)(=O)=O.OS(O)(=O)=O.OS(O)(=O)=O.OS(O)(=O)=O.OS(O)(=O)=O.CCN[C@@H]1C[C@H](N)[C@@H](O[C@H]2OC(CN)=CC[C@H]2N)[C@H](O)[C@H]1O[C@H]1OC[C@](C)(O)[C@H](NC)[C@H]1O.CCN[C@@H]1C[C@H](N)[C@@H](O[C@H]2OC(CN)=CC[C@H]2N)[C@H](O)[C@H]1O[C@H]1OC[C@](C)(O)[C@H](NC)[C@H]1O
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (4)
Quantity 
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (4)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (3)
  • (2)
  • (4)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)