missing translation for 'onlineSavingsMsg'
Learn More

Gilac™ Nesting Stacking Perforated Crate in Recycled HDPE

Nesting Stacking Perforated Crate in Recycled HDPE is stacking and nesting when half turned. Ideal for commercial kitchen use and the cold chain process.

Brand:  Gilac™ G180005

Additional Details : Weight : 2.00000kg

 View more versions of this product

Product Code. 17918441

  • £20.80 / Each
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • 600 x 400 perforated crate in recycled HDPE suitable for food contact.
  • Stacking and nesting when half turned.
  • Ideal for commercial kitchen use and the cold chain process.
  • Provides ventilation.
  • Large perforations meaning that temperatures can be brought down quickly and is easy to clean.
  • Recycled HDPE food crates offer the same level of professional quality as virgin HDPE crates.
    • Subject to very strict standards, recycled HDPE complies with all current food safety standards.
  • These new recycled HDPE crates also respond to changes in the anti-waste law.
  • By the end of 2022, purchasers from the State, local authorities and their groups will be obliged to acquire goods from reuse, or containing recycled materials.
Specifications

Specifications

9.8 gal.
Green
Rectangular
600 x 400 x 197 mm
23.6 in.
15.7 in.
7.8 in.
37 L
HDPE
Crate
600 mm
400 mm
197 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers