missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Nephronophthisis Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £386.00
Specifications
| Antigen | Nephronophthisis |
|---|---|
| Dilution | Western Blot : 2-10 μg/mL, Immunoprecipitation : 10-20 μg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30066860
|
Novus Biologicals™
NBP3-30792-250ug |
250 μg |
£386.00
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30066783
|
Novus Biologicals™
NBP3-30792-50ug |
50 μg |
£162.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nephronophthisis Polyclonal antibody specifically detects Nephronophthisis in Mouse samples. It is validated for Immunoprecipitation,Western BlotSpecifications
| Nephronophthisis | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Signal Transduction | |
| Phosphate buffered saline | |
| 4867 | |
| IgG | |
| Ammonium sulfate precipitation |
| Western Blot : 2-10 μg/mL, Immunoprecipitation : 10-20 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| FLJ97602, JBTS4NPH1Juvenile nephronophthisis 1 protein, nephrocystin 1, nephrocystin-1, nephronophthisis 1 (juvenile), SLSN1 | |
| Recombinant protein corresponding to the amino acid 612-691 of the C terminus region and having a molecular weight of 90 kD | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title