missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Nephronophthisis Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals™ NBP3-30792-250ug

 View more versions of this product

Product Code. 30066860

  • £386.00 / 250µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Nephronophthisis Polyclonal antibody specifically detects Nephronophthisis in Mouse samples. It is validated for Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Nephronophthisis
Polyclonal
Western Blot : 2-10 μg/mL, Immunoprecipitation : 10-20 μg/mL
FLJ97602, JBTS4NPH1Juvenile nephronophthisis 1 protein, nephrocystin 1, nephrocystin-1, nephronophthisis 1 (juvenile), SLSN1
Recombinant protein corresponding to the amino acid 612-691 of the C terminus region and having a molecular weight of 90 kD
250 μg
Cell Biology, Signal Transduction
4867
Store at -20°C. Avoid freeze/thaw cycles.
IgG
Immunoprecipitation, Western Blot
Unconjugated
Phosphate buffered saline
Rabbit
Ammonium sulfate precipitation
RUO
Primary
Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.